4-(Aminomethyl)benzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

283746-100G 283746-25G 283746-5G
SIAL283746-100GEA 174 EUR
SIAL283746-100G SIAL283746-25G SIAL283746-5G
4-(Aminomethyl)benzoesäure, Sigma-Aldrich®
4-(Aminomethyl)benzoic acid
Formel: C₈H₉NO₂
MDL: MFCD00010203
CAS-Nummer: 56-91-7
EINECS: 200-297-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR