N,N-Dimethylglycinethylester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Di-Me-Gly-OEt

M-Clarity™ Qualitätsstufe = MQ200

284394-25G
SIAL284394-25GEA 128 EUR
SIAL284394-25G
N,N-Dimethylglycinethylester, Sigma-Aldrich®
N,N-Dimethylglycinethylester
Formel: C₆H₁₃NO₂
Molecular Weight: 131,17 g/mol
MDL: MFCD00009175
CAS-Nummer: 33229-89-9
EINECS: 251-411-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR