D-Phenylalaninol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: D-Penylalaninol, D(+)-2-Amino-3-phenyl-1-propanol

M-Clarity™ Qualitätsstufe = MQ100

284491-5G
SIAL284491-5GEA 223 EUR
SIAL284491-5G
D-Phenylalaninol, Sigma-Aldrich®
D-Phenylalaninol
Formel: C₉H₁₃NO
Molecular Weight: 151,2 g/mol
MDL: MFCD00064298
CAS-Nummer: 5267-64-1
EINECS: 226-086-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR