3-Indolcarbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1H-Indole-3-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

284734-1G 284734-5G
SIAL284734-1GEA 26 EUR
SIAL284734-1G SIAL284734-5G
3-Indolcarbonsäure, Sigma-Aldrich®
3-Indolcarbonsäure
Formel: C₉H₇NO₂
MDL: MFCD00005624
CAS-Nummer: 771-50-6
EINECS: 212-231-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR