6-Methylnicotinamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Methylpyridine-3-carboxamide

M-Clarity™ Qualitätsstufe = MQ100

284769-5G
SIAL284769-5GEA 131 EUR
SIAL284769-5G
6-Methylnicotinamid, Sigma-Aldrich®
6-Methylnicotinamid
Formel: C₇H₈N₂O
Molecular Weight: 136,15 g/mol
MDL: MFCD00006342
CAS-Nummer: 6960-22-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR