Methyl-6-methylnicotinat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 6-methylnicotinate

M-Clarity™ Qualitätsstufe = MQ100

284777-10G
SIAL284777-10GEA 153 EUR
SIAL284777-10G
Methyl-6-methylnicotinat, Sigma-Aldrich®
Methyl-6-methylnicotinat
Formel: C₈H₉NO₂
Molecular Weight: 151,16 g/mol
MDL: MFCD00006340
CAS-Nummer: 5470-70-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR