trans-1,2-Cyclobutandicarbonsäure, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
28684-1G
28684-5G
SIAL28684-1GEA
196
EUR
SIAL28684-1G
SIAL28684-5G
trans-1,2-Cyclobutandicarbonsäure, Sigma-Aldrich®
trans-1,2-Cyclobutandicarbonsäure
Formel:
C₆H₈O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00068260 CAS-Nummer: 1124-13-6 EINECS: 214-392-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...