Cyclopentadienylmangantricarbonyl, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Cymantrene, Cyclopentadienylmanganese(I) tricarbonyl

M-Clarity™ Qualitätsstufe = MQ100

288055-1G
SIAL288055-1GEA 87.9 EUR
SIAL288055-1G
Cyclopentadienylmangantricarbonyl, Sigma-Aldrich®
Cyclopentadienylmangantricarbonyl
Formel: C₈H₅MnO₃
Molecular Weight: 204,06 g/mol
MDL: MFCD00001440
CAS-Nummer: 12079-65-1
EINECS: 235-142-4
UN: 3466
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR