(Triethylphosphin)gold(I)chlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Chlorotriethylphosphinegold(I)

M-Clarity™ Qualitätsstufe = MQ100

288225-1G 288225-250MG
SIAL288225-1GEA 340 EUR
SIAL288225-1G SIAL288225-250MG
(Triethylphosphin)gold(I)chlorid, Sigma-Aldrich®
(Triethylphosphine)gold(I) chloride
Formel: C₆H₁₅AuClP
Molecular Weight: 350.577 g/mol
MDL: MFCD00013205
CAS-Nummer: 15529-90-5
EINECS: 239-572-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR