Indium(III)oxid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Diindiumtrioxid, Indiumtrioxid
MQ100
289418-10G
289418-50G
SIAL289418-10GEA
157
EUR
SIAL289418-10G
SIAL289418-50G
Indium(III)oxid, Sigma-Aldrich®
Indium(III)oxid
Formel:
In₂O₃ Molecular Weight: 277.64 g/mol Schmelzpunkt: 715.05 Dichte: 7.18 g/cm³ (20 °C) |
MDL:
MFCD00011060 CAS-Nummer: 1312-43-2 EINECS: 215-193-9 Merck Index: 12,04984 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...