Kaliumbenzoat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Benzoic acid potassium salt
MQ100
290009-1KG
290009-250G
SIAL290009-1KGEA
117
EUR
SIAL290009-1KG
SIAL290009-250G
Kaliumbenzoat, Sigma-Aldrich®
Kaliumbenzoat
Formel:
C₇H₅KO₂ Molecular Weight: 160.21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013061 CAS-Nummer: 582-25-2 EINECS: 209-481-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...