2-Amino-5-nitro-4-picolin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Methyl-5-nitro-2-pyridylamin, 2-Amino-4-methyl-5-nitropyridin

M-Clarity™ Qualitätsstufe = MQ100

290092-1G
SIAL290092-1GEA 32.2 EUR
SIAL290092-1G
2-Amino-5-nitro-4-picolin, Sigma-Aldrich®
2-Amino-5-nitro-4-picolin
Formel: C₆H₇N₃O₂
Molecular Weight: 153,14 g/mol
Schmelzpunkt: 220 °C
MDL: MFCD00010692
CAS-Nummer: 21901-40-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR