2-Hydroxy-5-nitro-4-picolin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Methyl-5-nitro-2-pyridon, 2-Hydroxy-4-methyl-5-nitropyridin

M-Clarity™ Qualitätsstufe = MQ100

290106-5G
SIAL290106-5GEA 154 EUR
SIAL290106-5G
2-Hydroxy-5-nitro-4-picolin, Sigma-Aldrich®
2-Hydroxy-5-nitro-4-picolin
Formel: C₆H₆N₂O₃
Molecular Weight: 154,13 g/mol
Schmelzpunkt: 189…190 °C
MDL: MFCD00010690
CAS-Nummer: 21901-41-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR