2-Hydroxy-3-nitro-4-picolin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Hydroxy-4-methyl-3-nitropyridin, 4-Methyl-3-nitro-2-pyridon

M-Clarity™ Qualitätsstufe = MQ100

290114-5G
SIAL290114-5GEA 165 EUR
SIAL290114-5G
2-Hydroxy-3-nitro-4-picolin, Sigma-Aldrich®
2-Hydroxy-3-nitro-4-picolin
Formel: C₆H₆N₂O₃
Molecular Weight: 154,13 g/mol
Schmelzpunkt: 234…235 °C
MDL: MFCD00010689
CAS-Nummer: 21901-18-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR