3,5-Difluorbenzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

290203-5G
SIAL290203-5GEA 25.5 EUR
SIAL290203-5G
3,5-Difluorbenzonitril, Sigma-Aldrich®
3,5-Difluorbenzonitril
Formel: C₇H₃F₂N
Molecular Weight: 139,1 g/mol
Siedepunkt: 159…160 °C (1013 hPa)
MDL: MFCD00010311
CAS-Nummer: 64248-63-1
EINECS: 264-752-3
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR