trans-2,4-Difluorzimtsäure, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 2,4-Difluorcinnamsäure, 3-(2,4-Difluorphenyl)propensäure

M-Clarity™ Qualitätsstufe = MQ200

290335-5G
SIAL290335-5GEA 128 EUR
SIAL290335-5G
trans-2,4-Difluorzimtsäure, Sigma-Aldrich®
trans-2,4-Difluorzimtsäure
Formel: C₉H₆F₂O₂
Molecular Weight: 184,14 g/mol
MDL: MFCD00010317
CAS-Nummer: 94977-52-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR