trans-2,5-Difluorzimtsäure, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
3-(2,5-Difluorphenyl)propensäure, 2,5-Difluorcinnamsäure
M-Clarity™ Qualitätsstufe = MQ100
290343-5G
SIAL290343-5GEA
105
EUR
SIAL290343-5G
trans-2,5-Difluorzimtsäure, Sigma-Aldrich®
trans-2,5-Difluorzimtsäure
Formel:
C₉H₆F₂O₂ Molecular Weight: 184,14 g/mol |
MDL:
MFCD00010318 CAS-Nummer: 112898-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...