trans-3,4-Difluorzimtsäure, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 3-(3,4-Difluorphenyl)propensäure, 3,4-Difluorcinnamsäure

M-Clarity™ Qualitätsstufe = MQ100

290378-5G
SIAL290378-5GEA 30.6 EUR
SIAL290378-5G
trans-3,4-Difluorzimtsäure, Sigma-Aldrich®
trans-3,4-Difluorzimtsäure
Formel: C₉H₆F₂O₂
Molecular Weight: 184,14 g/mol
MDL: MFCD00010320
CAS-Nummer: 112897-97-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR