(2,3-Difluorphenyl)essigsäure, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 2-(2,3-Difluorophenyl)acetic acid

M-Clarity™ Qualitätsstufe = MQ100

290416-1G
SIAL290416-1GEA 52.8 EUR
SIAL290416-1G
(2,3-Difluorphenyl)essigsäure, Sigma-Aldrich®
(2,3-Difluorphenyl)essigsäure
Formel: (C₆H₃F₂)CH₂COOH
Molecular Weight: 172,13 g/mol
MDL: MFCD00040968
CAS-Nummer: 145689-41-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR