trans-3-Fluorzimtsäure, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: trans-3-Fluorcinnamsäure, 3-(3-Fluorphenyl)propensäure, 3-(m-Fluorphenyl)propensäure

M-Clarity™ Qualitätsstufe = MQ100

290483-5G
SIAL290483-5GEA 41.8 EUR
SIAL290483-5G
trans-3-Fluorzimtsäure, Sigma-Aldrich®
trans-3-Fluorzimtsäure
Formel: C₉H₇FO₂
Molecular Weight: 166,15 g/mol
MDL: MFCD00004383
CAS-Nummer: 20595-30-6
EINECS: 243-902-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR