Tris(pentafluorphenyl)phosphin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Perfluortriphenylphosphin

M-Clarity™ Qualitätsstufe = MQ200

290572-1G
SIAL290572-1GEA 67.7 EUR
SIAL290572-1G
Tris(pentafluorphenyl)phosphin, Sigma-Aldrich®
Tris(pentafluorphenyl)phosphin
Formel: C₁₈F₁₅P
Molecular Weight: 532,15 g/mol
MDL: MFCD00079654
CAS-Nummer: 1259-35-4
EINECS: 215-021-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR