4'-Phenoxyacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: p-Phenoxyacetophenon, 4-Phenoxy-1-acetylbenzol, 1-(4-Phenoxyphenyl)ethanon

290742-5G
SIAL290742-5GEA 99.6 EUR
SIAL290742-5G
4'-Phenoxyacetophenon, Sigma-Aldrich®
4'-Phenoxyacetophenon
Formel: C₁₄H₁₂O₂
Molecular Weight: 212,25 g/mol
Siedepunkt: 119…120 °C (0,03 mmHg)
Schmelzpunkt: 56 °C
MDL: MFCD00008744
CAS-Nummer: 5031-78-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR