2-Ethylhexylmethacrylat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

290807-1L 290807-25ML
SIAL290807-1LEA 227 EUR
SIAL290807-1L SIAL290807-25ML
2-Ethylhexylmethacrylat, Sigma-Aldrich®
2-Ethylhexylmethacrylat
Formel: C₁₂H₂₂O₂
MDL: MFCD00009494
CAS-Nummer: 688-84-6
EINECS: 211-708-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR