α,α,α,α',α',α'-Hexafluor-p-xylol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: α,α,α,β,β,β-Hexafluor-p-xylol

M-Clarity™ Qualitätsstufe = MQ100

290912-25G
SIAL290912-25GEA 56.5 EUR
SIAL290912-25G
α,α,α,α',α',α'-Hexafluor-p-xylol, Sigma-Aldrich®
α,α,α,α',α',α'-Hexafluor-p-xylol
Formel: C₈H₄F₆
Molecular Weight: 214,11 g/mol
Siedepunkt: 113…115 °C (1013 hPa)
Schmelzpunkt: 3 °C
Dichte: 1,3808 g/cm³ (25 °C)
MDL: MFCD00000402
CAS-Nummer: 433-19-2
EINECS: 207-086-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR