Piperonylbutoxid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
5-[2-(2-Butoxyethoxy)ethoxymethyl]-6-propyl-1,3-benzodioxole, 5-((2-(2-Butoxyethoxy)ethoxy)methyl)-6-propylbenzo[d][1,3]dioxole, NSC 8401
M-Clarity™ Qualitätsstufe = MQ200
291102-100ML
SIAL291102-100MLEA
76.3
EUR
SIAL291102-100ML
Piperonylbutoxid, Sigma-Aldrich®
Piperonylbutoxid
Formel:
C₁₉H₃₀O₅ Molecular Weight: 338,44 g/mol |
MDL:
MFCD00005842 CAS-Nummer: 51-03-6 EINECS: 200-076-7 UN: 3082 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...