4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosan ([2.2.1]Cryptand, Kryptofix® 221), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Kryptofix® 221, 1,10-Diaza-4,7,13,16,21-pentaoxabicyclo[8.8.5]tricosan, [2.2.1]Cryptand

M-Clarity™ Qualitätsstufe = MQ200

291161-1G
SIAL291161-1GEA 492 EUR
SIAL291161-1G
4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosan ([2.2.1]Cryptand, Kryptofix® 221), Sigma-Aldrich®
4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosan ([2.2.1]Cryptand, Kryptofix® 221)
Formel: C₁₆H₃₂N₂O₅
Molecular Weight: 332,44 g/mol
Dichte: 1,11 g/cm³ (20 °C)
MDL: MFCD00005108
CAS-Nummer: 31364-42-8
EINECS: 250-592-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR