p-Tolylacetat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Acetic acid p-Cresyl ester, Acetic acid p-Tolyl ester, p-Tolyl Acetate, p-Cresyl acetate

M-Clarity™ Qualitätsstufe = MQ100

291390-250ML
SIAL291390-250MLEA 126 EUR
SIAL291390-250ML
p-Tolylacetat, Sigma-Aldrich®
p-Tolylacetat
Formel: C₉H₁₀O₂
Molecular Weight: 150,1745 g/mol
MDL: MFCD00008703
CAS-Nummer: 140-39-6
EINECS: 205-413-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR