Bortrifluorid-Ethylamin-Komplex, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
292249-100G
292249-500G
SIAL292249-100GEA
96
EUR
SIAL292249-100G
SIAL292249-500G
Bortrifluorid-Ethylamin-Komplex, Sigma-Aldrich®
Bortrifluorid-Ethylamin-Komplex
Formel:
C₂H₇BF₃N |
MDL:
MFCD00144277 CAS-Nummer: 75-23-0 EINECS: 200-852-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...