α-Bromstyrol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: alfa-Bromostyrene, α-Bromostyrene, alpha-Bromostyrene

MQ100

292273-5G
SIAL292273-5GEA 69.8 EUR
SIAL292273-5G
α-Bromstyrol, Sigma-Aldrich®
α-Bromstyrol
Formel: C₈H₇Br
Molecular Weight: 183.0452 g/mol
MDL: MFCD00012229
CAS-Nummer: 98-81-7
EINECS: 202-702-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR