5-Chlor-2-methoxybenzoesäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Chlor-o-anissäure

M-Clarity™ Qualitätsstufe = MQ100

292494-25G
SIAL292494-25GEA 38 EUR
SIAL292494-25G
5-Chlor-2-methoxybenzoesäure, Sigma-Aldrich®
5-Chlor-2-methoxybenzoesäure
Formel: C₈H₇ClO₃
Molecular Weight: 186,59 g/mol
MDL: MFCD00017546
CAS-Nummer: 3438-16-2
EINECS: 222-343-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR