Cyclohexen-1-carbonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Cyclohexene-1-carbonitrile, Cyclohex-1-enecarbonitrile

29252-10ML
SIAL29252-10MLEA 295 EUR
SIAL29252-10ML
Cyclohexen-1-carbonitril, Sigma-Aldrich®
Cyclohexen-1-carbonitril
Formel: C₇H₉N
Molecular Weight: 107,16 g/mol
MDL: MFCD00013772
CAS-Nummer: 1855-63-6
EINECS: 217-454-2
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR