2-Hydroxyethylacrylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Acrylsäure(2-hydroxyethyl)ester, Propensäure(2-hydroxyethyl)ester

M-Clarity™ Qualitätsstufe = MQ100

292818-1L 292818-250ML 292818-18L
SIAL292818-1LEA 262 EUR
SIAL292818-1L SIAL292818-250ML SIAL292818-18L
2-Hydroxyethylacrylat, Sigma-Aldrich®
2-Hydroxyethylacrylat
Formel: C₅H₈O₃
Molecular Weight: 116.12 g/mol
MDL: MFCD00002865
CAS-Nummer: 818-61-1
EINECS: 212-454-9
UN: 2927
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR