Acetylchlorid-[2-13C] (99% ¹³C ), Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ200
293148-250MG
SIAL293148-250MGEA
322
EUR
SIAL293148-250MG
Acetylchlorid-[2-13C] (99% ¹³C ), Sigma-Aldrich®
Acetylchlorid-[2-13C]
Formel:
C₂H₃ClO |
CAS-Nummer:
14770-40-2 UN: 1717 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...