CAPS (3-(Cyclohexylamino)propansulfonsäure), Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Cyclohexylaminopropanesulphonic acid

29337-100G
SIAL29337-100GEA 508 EUR
SIAL29337-100G
CAPS (3-(Cyclohexylamino)propansulfonsäure), Sigma-Aldrich®
CAPS (3-(Cyclohexylamino)propansulfonsäure)
Formel: C₆H₁₁NH(CH₂)₃SO₃H
Molecular Weight: 221,32 g/mol
Schmelzpunkt: 324 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003837
CAS-Nummer: 1135-40-6
EINECS: 214-492-1
Merck Index: 13,01773

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR