2-Chinoxalincarbonsäure, Sigma-Aldrich®

Lieferant: Merck

293407-500MG
SIAL293407-500MGEA 108 EUR
SIAL293407-500MG
2-Chinoxalincarbonsäure, Sigma-Aldrich®
2-Quinoxalinecarboxylic acid
Formel: C₉H₆N₂O₂
Molecular Weight: 174,16 g/mol
MDL: MFCD00012334
CAS-Nummer: 879-65-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR