1-Methyluracil, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,4-Dihydroxy-1-methylpyrimidin

M-Clarity™ Qualitätsstufe = MQ200

293768-500MG
SIAL293768-500MGEA 236 EUR
SIAL293768-500MG
1-Methyluracil, Sigma-Aldrich®
1-Methyluracil
Formel: C₅H₆N₂O₂
Molecular Weight: 126.11 g/mol
MDL: MFCD00038666
CAS-Nummer: 615-77-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR