cis-2,3-Epoxybutan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

294047-1G
SIAL294047-1GEA 52.4 EUR
SIAL294047-1G
cis-2,3-Epoxybutan, Sigma-Aldrich®
cis-2,3-Epoxybutane
Formel: C₄H₈O
Molecular Weight: 72,11 g/mol
MDL: MFCD00005128
CAS-Nummer: 1758-33-4
UN: 3271
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR