Kaliumperfluor-1-butansulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Nonafluoro-1-butanesulphonic acid potassium salt, Potassium Perfluoro-1-butanesulphonate, Potassium Nonafluoro-1-butanesulphonate, Perfluoro-1-butanesulphonic acid potassium salt, Potassium nonafluoro-1-butanesulphonate

M-Clarity™ Qualitätsstufe = MQ100

294209-10G
SIAL294209-10GEA 61.1 EUR
SIAL294209-10G
Kaliumperfluor-1-butansulfonat, Sigma-Aldrich®
Kaliumperfluor-1-butansulfonat
Formel: C₄F₉KO₃S
Molecular Weight: 338,1899 g/mol
MDL: MFCD00007515
CAS-Nummer: 29420-49-3
EINECS: 249-616-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR