4-Iod-D-phenylalanin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: H-D-Phe(4-I)-OH

M-Clarity™ Qualitätsstufe = MQ100

294462-500MG
SIAL294462-500MGEA 252 EUR
SIAL294462-500MG
4-Iod-D-phenylalanin, Sigma-Aldrich®
4-Iod-D-phenylalanin
Formel: C₉H₁₀INO₂
Molecular Weight: 291,1 g/mol
MDL: MFCD00063067
CAS-Nummer: 62561-75-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR