(±)-1-(1-Naphthyl)ethylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(Naphthalen-1-yl)ethanamine

M-Clarity™ Qualitätsstufe = MQ100

294926-5G
SIAL294926-5GEA 71.1 EUR
SIAL294926-5G
(±)-1-(1-Naphthyl)ethylamin, Sigma-Aldrich®
(±)-1-(1-Naphthyl)ethylamin
Formel: C₁₂H₁₃N
Molecular Weight: 171,24 g/mol
MDL: MFCD00004014
CAS-Nummer: 42882-31-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR