2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Panthenol, DL-Pantothenol, 2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide

M-Clarity™ Qualitätsstufe = MQ200

295787-100G
SIAL295787-100GEA 100 EUR
SIAL295787-100G
2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamid, Sigma-Aldrich®
DL-Panthenol
Formel: C₉H₁₉NO₄
Molecular Weight: 205.25 g/mol
MDL: MFCD00002944
CAS-Nummer: 16485-10-2
EINECS: 240-540-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR