1,10-Diaza-18-krone-6 (1,4,10,13-Tetraoxa-7,16-diazacyclooctadecan, Kryptofix® 22), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

295809-1G
SIAL295809-1GEA 236 EUR
SIAL295809-1G
1,10-Diaza-18-krone-6 (1,4,10,13-Tetraoxa-7,16-diazacyclooctadecan, Kryptofix® 22), Sigma-Aldrich®
1,10-Diaza-18-krone-6 (1,4,10,13-Tetraoxa-7,16-diazacyclooctadecan, Kryptofix® 22)
Formel: C₁₂H₂₆N₂O₄
MDL: MFCD00005112
CAS-Nummer: 23978-55-4
EINECS: 245-965-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR