N,N-Bis(trifluormethansulfonyl)anilin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,1,1-Trifluoro-N-phenyl-N-((trifluoromethyl)sulphonyl)methanesulphonamide, N-Phenylbis(trifluoromethanesulphonimide)

M-Clarity™ Qualitätsstufe = MQ200

295973-25G 295973-5G
SIAL295973-25GEA 190 EUR
SIAL295973-25G SIAL295973-5G
N,N-Bis(trifluormethansulfonyl)anilin, Sigma-Aldrich®
N,N-Bis(trifluormethansulfonyl)anilin
Formel: C₈H₅F₆NO₄S₂
MDL: MFCD00000404
CAS-Nummer: 37595-74-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR