N,N-Bis(trifluormethansulfonyl)anilin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1,1,1-Trifluoro-N-phenyl-N-((trifluoromethyl)sulphonyl)methanesulphonamide, N-Phenylbis(trifluoromethanesulphonimide)
M-Clarity™ Qualitätsstufe = MQ200
295973-25G
295973-5G
SIAL295973-25GEA
190
EUR
SIAL295973-25G
SIAL295973-5G
N,N-Bis(trifluormethansulfonyl)anilin, Sigma-Aldrich®
N,N-Bis(trifluormethansulfonyl)anilin
Formel:
C₈H₅F₆NO₄S₂ |
MDL:
MFCD00000404 CAS-Nummer: 37595-74-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...