Acetonitril-[2-13C,D3] (99% ¹³C , 99,5% D), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Acetonitrile-[2-¹³C,²H₃]

M-Clarity™ Qualitätsstufe = MQ200

296015-1G
SIAL296015-1GEA 3700 EUR
SIAL296015-1G
Acetonitril-[2-13C,D3] (99% ¹³C , 99,5% D), Sigma-Aldrich®
Acetonitril-[2-13C,D3]
Formel: C₂D₃N
MDL: MFCD00001880
CAS-Nummer: 62663-01-8
UN: 1648
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR