2-Amino-3-nitrophenol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 2-Hydroxy-6-nitroanilin

M-Clarity™ Qualitätsstufe = MQ100

297003-5G
SIAL297003-5GEA 26 EUR
SIAL297003-5G
2-Amino-3-nitrophenol, Sigma-Aldrich®
2-Amino-3-nitrophenol
Formel: C₆H₆N₂O₃
Molecular Weight: 154.13 g/mol
MDL: MFCD00010875
CAS-Nummer: 603-85-0
EINECS: 210-060-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR