(R)-(-)-2-Amino-1-propanol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
D(-)-Alaninol, D(-)-2-Amino-1-propanol
M-Clarity™ Qualitätsstufe = MQ100
297682-1G
SIAL297682-1GEA
40.8
EUR
SIAL297682-1G
(R)-(-)-2-Amino-1-propanol, Sigma-Aldrich®
(R)-(-)-2-Amino-1-propanol
Formel:
C₃H₉NO Molecular Weight: 75.11 g/mol Siedepunkt: 173 Dichte: 0.96 g/cm³ (20 °C) |
MDL:
MFCD00064413 CAS-Nummer: 35320-23-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...