2,2,2-Trifluorethylacrylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Acrylsäure(2,2,2-trifluorethyl)ester, Propensäure(2,2,2-trifluorethyl)ester

M-Clarity™ Qualitätsstufe = MQ200

297720-25G 297720-5G
SIAL297720-25GEA 161 EUR
SIAL297720-25G SIAL297720-5G
2,2,2-Trifluorethylacrylat, Sigma-Aldrich®
2,2,2-Trifluorethylacrylat
Formel: C₅H₅F₃O₂
Molecular Weight: 154.09 g/mol
MDL: MFCD00000444
CAS-Nummer: 407-47-6
EINECS: 206-987-6
UN: 3272
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR