2-Chlor-4-fluorbenzoesäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
297836-25G
SIAL297836-25GEA
79.5
EUR
SIAL297836-25G
2-Chlor-4-fluorbenzoesäure, Sigma-Aldrich®
2-Chloro-4-fluorobenzoic acid
Formel:
C₇H₄ClFO₂ Molecular Weight: 174,56 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010615 CAS-Nummer: 2252-51-9 EINECS: 218-845-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...