Ethyl-p-tolylacetat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(4-Methylphenyl)essigsäureethylester, p-Tolylessigsäureethylester
M-Clarity™ Qualitätsstufe = MQ100
297852-25G
SIAL297852-25GEA
112
EUR
SIAL297852-25G
Ethyl-p-tolylacetat, Sigma-Aldrich®
Ethyl-p-tolylacetat
Formel:
(C₆H₄(CH₃))CH₂COOCH₂CH₃ Molecular Weight: 178,23 g/mol Siedepunkt: 116 °C (1013 hPa) Dichte: 1 g/cm³ (20 °C) |
MDL:
MFCD00009181 CAS-Nummer: 14062-19-2 EINECS: 237-904-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...